Diethylentriamin 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2,2'-Iminodiethylamin, Bis(2-aminoethyl)amin, 2,2'-Diaminodiethylamin, 2,2'-Iminodi(ethylamin), 3-Azapentan-1,5-diamin

A11033.0F A11033.AE A11033.AP
A11033.0FEA 126 EUR
A11033.0F A11033.AE A11033.AP
Diethylentriamin 99%
Diethylentriamin

Purity: 99% CAS No.: 111-40-0 Molecular Formula: C4H13N3 Formula Weight: 103.17 Flash Point: 94°(201°F) Refractive Index: 1.4830 Odour: Ammonia-like Melting Point: -40° Boiling Point: 200-204° Beilstein Reference: 605314 UN No.: 2079 MDL No.: MFCD00008171

Formel: C₄H₁₃N₃
Molecular Weight: 103,17 g/mol
Siedepunkt: 200…204 °C
Schmelzpunkt: –40 °C
Dichte: 0,952
Flammpunkt: 94 °C (201 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008171
CAS-Nummer: 111-40-0
EINECS: 203-865-4
UN: 2079
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR