Acetonoxim ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Propan-2-one oxime, Acetoxime, Acetone oxime, 2-Propanone oxime

A10802.0B A10802.18 A10802.30
A10802.0BEA 230 EUR
A10802.0B A10802.18 A10802.30
Acetonoxim ≥98%
Acetonoxim

Purity: 98% CAS No.: 127-06-0 Molecular Formula: C3H7NO Formula Weight: 73.10 Flash Point: 60°(140°F) Melting Point: 60-63° Boiling Point: 133-135° Beilstein Reference: 1560146 Merck Reference: 14,75 UN No.: 1325 MDL No.: MFCD00002118

Formel: C₃H₇NO
Molecular Weight: 73,09 g/mol
Siedepunkt: 133…135 °C
Schmelzpunkt: 60…63 °C
Dichte: 0,901
Flammpunkt: 60 °C (140 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002118
CAS-Nummer: 127-06-0
EINECS: 204-820-1
UN: 1325
ADR: 4.1,III
Merck Index: 14,75

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR