Dimethylacetal 98
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Acetaldehyddimethylacetal, 1,1-Dimethoxyethan
A10939.14
A10939.22
A10939.36
A10939.14EA
35.8
EUR
A10939.14
A10939.22
A10939.36
Dimethylacetal 98
Dimethylacetal
Purity: 98+%
CAS No.: 534-15-6
Molecular Formula: CH3CH(OCH3)2
Formula Weight: 90.12
Flash Point: -17°(1°F)
Refractive Index: 1.3670
Odour: Sweet
Melting Point: -113°
Boiling Point: 63-65°
Beilstein Reference: 1697039
Merck Reference: 14,3226
UN No.: 2377
MDL No.: MFCD00008493
Formel:
C₄H₁₀O₂ Molecular Weight: 90.12 g/mol Siedepunkt: 63…65 °C Schmelzpunkt: –113 °C Dichte: 0.852 Flammpunkt: –17 °C (1 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008493 CAS-Nummer: 534-15-6 EINECS: 208-589-8 UN: 2377 ADR: 3 II Merck Index: 13,03253 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...