1-Pentansulfonsäure Natriumsalz
Lieferant: FLUOROCHEM
Synonyme:
Natriumpentylsulfonat
008051-25G
008051-10G
008051-100G
FLUO008051-25GEA
77
EUR
FLUO008051-25G
FLUO008051-10G
FLUO008051-100G
1-Pentansulfonsäure Natriumsalz
1-Pentanesulphonic acid sodium salt
Formel:
C₅H₁₁O₃SNa Molecular Weight: 174.2 g/mol Schmelzpunkt: > 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007541 CAS-Nummer: 22767-49-3 EINECS: 245-208-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...