Dimethylitaconat ≥97%

Lieferant: Thermo Scientific

Synonyme: Itaconic acid dimethyl ester, Methylenesuccinic acid dimethyl ester

L12652.22 L12652.36 AB179531.0500
L12652.22EA 45.8 EUR
L12652.22 L12652.36 ABCRAB179531.0500
Dimethylitaconat ≥97%
Dimethylitaconat

Purity: 97% CAS No.: 617-52-7 Molecular Formula: C7H10O4 Formula Weight: 158.16 Flash Point: 100°(212°F) Melting Point: 33-39° Boiling Point: 207-208° Beilstein Reference: 386674 MDL No.: MFCD00008446

Formel: C₇H₁₀O₄
Molecular Weight: 158,15 g/mol
Siedepunkt: 207…208 °C
Schmelzpunkt: 33…39 °C
Dichte: 1,124
Flammpunkt: 100 °C (212 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008446
CAS-Nummer: 617-52-7
EINECS: 210-519-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR