Dimethylitaconat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Itaconic acid dimethyl ester, Methylenesuccinic acid dimethyl ester

592498-100G 592498-25G
SIAL592498-100GEA 413 EUR
SIAL592498-100G SIAL592498-25G
Dimethylitaconat, Sigma-Aldrich®
Dimethylitaconat
Formel: C₇H₁₀O₄
Molecular Weight: 158,15 g/mol
Siedepunkt: 208 °C (1013 hPa)
Schmelzpunkt: 38 °C
Dichte: 1,1399 g/cm³ (14.7 °C)
MDL: MFCD00008446
CAS-Nummer: 617-52-7
EINECS: 210-519-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR