Ethyl-2-bromisobutyrat 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
α-Bromisobuttersäureethylester, Ethyl-2-brom-2-methylpropionat, 2-Bromisobuttersäureethylester, 2-Brom-2-methylpropionsäureethylester
118130050
118135000
ACRO118130050EA
21.2
EUR
ACRO118130050
ACRO118135000
Ethyl-2-bromisobutyrat 98%
Ethyl-2-bromisobutyrat
Formel:
C₆H₁₁BrO₂ Molecular Weight: 195.06 g/mol Siedepunkt: 65 Dichte: 1.329 g/cm³ Flammpunkt: 60 °C |
MDL:
MFCD00000123 CAS-Nummer: 600-00-0 EINECS: 209-980-6 UN: 1993 |
Spezifikation Testergebnisse
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4430 to 1.4460 (20°C, 589 nm) |
GC | ≥97.5 % |
Sodium (Na) | 30.5 to 32.5 % (as Na2SO4) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...