Ethylisobutyrat ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Isobuttersäureethylester
A12966.0F
A12966.AE
A12966.AP
A12966.0FEA
214
EUR
A12966.0F
A12966.AE
A12966.AP
Ethylisobutyrat ≥98%
Ethylisobutyrat
Purity: 98%
CAS No.: 97-62-1
Molecular Formula: (CH3)2CHCO2CH2CH3
Formula Weight: 116.16
Flash Point: 13°(55°F)
Refractive Index: 1.3870
Odour: Pleasant
Melting Point: -88°
Boiling Point: 108-110°
Beilstein Reference: 773846
Merck Reference: 14,3814
UN No.: 2385
MDL No.: MFCD00009165
Formel:
C₆H₁₂O₂ Molecular Weight: 116,16 g/mol Siedepunkt: 108…110 °C Schmelzpunkt: –88 °C Dichte: 0,869 Flammpunkt: 13 °C (55 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009165 CAS-Nummer: 97-62-1 EINECS: 202-595-4 UN: 2385 ADR: 3,II Merck Index: 14,3814 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...