Calcium-2-ethylhexanoat ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Ethylhexansäure Calciumsalz, Heptan-3-carbonsäure Calciumsalz, 2-Ethylcapronsäure Calciumsalz

A18266.06 A18266.14 A18266.22
A18266.06EA 46.2 EUR
A18266.06 A18266.14 A18266.22
Calcium-2-ethylhexanoat ≥98%
Calcium-2-ethylhexanoat

Purity: 98% CAS No.: 136-51-6 Molecular Formula: C16H30CaO4 Formula Weight: 326.50 Odour: Odourless

Formel: C₁₆H₃₀CaO₄
Molecular Weight: 326,49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014001
CAS-Nummer: 136-51-6
EINECS: 205-249-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR