Zinn(II)-2-ethylhexanoat, tech.

Lieferant: Thermo Scientific
Achtung

Synonyme: Zinnbis(2-ethylhexanoat), Heptan-3-carbonsäure Zinn(II)salz, 2-Ethylhexansäure Zinn(II)salz, 2-Ethylcapronsäure Zinn(II)salz

18590.22 18590.36 18590.A3 AB204036.0100
18590.22EA 45 EUR
18590.22 18590.36 18590.A3 ABCRAB204036.0100
Zinn(II)-2-ethylhexanoat, tech.
Zinn(II)-2-ethylhexanoat

CAS No.: 301-10-0 Molecular Formula: C16H30O4Sn Formula Weight: 405.11 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.251 Refractive Index: 1.4940 Appearance: Colourless to pale yellow MDL No.: MFCD00002676

Formel: C₁₆H₃₀O₄Sn
Molecular Weight: 405,12 g/mol
Dichte: 1,251
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002676
CAS-Nummer: 301-10-0
EINECS: 206-108-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR