Zinn(II)-2-ethylhexanoat 95% (Sn-Basis)

Lieferant: Thermo Scientific
Achtung

Synonyme: Zinnbis(2-ethylhexanoat), Heptan-3-carbonsäure Zinn(II)salz, 2-Ethylhexansäure Zinn(II)salz, 2-Ethylcapronsäure Zinn(II)salz

B23612.14 B23612.22
B23612.14EA 30 EUR
B23612.14 B23612.22
Zinn(II)-2-ethylhexanoat 95% (Sn-Basis)
Zinn(II)-2-ethylhexanoat

Purity: 96% CAS No.: 301-10-0 Molecular Formula: C16H30O4Sn Formula Weight: 405.11 Flash Point: 110°(230°F) Refractive Index: 1.4940 Odour: Characteristic MDL No.: MFCD00002676

Formel: C₁₆H₃₀O₄Sn
Molecular Weight: 405,12 g/mol
Dichte: 1,251
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002676
CAS-Nummer: 301-10-0
EINECS: 206-108-6

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Clear colorless to yellow
Form Viscous liquid
Assay from Supplier's CofA ≥94.0 to ≤106.0% (based on Sn EA)
Elemental Analysis: Carbon (C) 44.59-50.28%
Elemental Analysis: Tin (Sn) 27.55-31.06%
Identification (FTIR) Conforms
Refractive Index 1.4910-1.4970 @ 20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR