Zinn(II)-2-ethylhexanoat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Zinnbis(2-ethylhexanoat), Heptan-3-carbonsäure Zinn(II)salz, 2-Ethylhexansäure Zinn(II)salz, 2-Ethylcapronsäure Zinn(II)salz
S3252-1KG
S3252-100G
S3252-250G
SIALS3252-1KGEA
180
EUR
SIALS3252-1KG
SIALS3252-100G
SIALS3252-250G
Zinn(II)-2-ethylhexanoat, Sigma-Aldrich®
Zinn(II)-2-ethylhexanoat
Formel:
C₁₆H₃₀O₄Sn Molecular Weight: 405,12 g/mol |
MDL:
MFCD00002676 CAS-Nummer: 301-10-0 EINECS: 206-108-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...