Pinakol ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Tetramethylethylenglycol, Pinakon, 2,3-Dimethyl-2,3-butandiol

B25006.14 B25006.22 B25006.36
B25006.14EA 47 EUR
B25006.14 B25006.22 B25006.36
Pinakol ≥99%
Pinakol

Purity: 99% CAS No.: 76-09-5 Molecular Formula: C6H14O2 Formula Weight: 118.18 Flash Point: 77°(170°F) Melting Point: 42-45° Boiling Point: 171-172° Beilstein Reference: 1340501 Merck Reference: 14,7439 MDL No.: MFCD00004462

Formel: C₆H₁₄O₂
Molecular Weight: 118,18 g/mol
Siedepunkt: 171…172 °C
Schmelzpunkt: 42…45 °C
Dichte: 0,967 g/cm³ (20 °C)
Flammpunkt: 77 °C (170 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004462
CAS-Nummer: 76-09-5
EINECS: 200-933-5
Merck Index: 14,7439

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR