Benzylbutyrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Butyric acid benzyl ester, Butanoic acid benzyl ester
M-Clarity™ Qualitätsstufe = MQ300
W214000-10KG-K
W214000-1KG-K
W214000-5KG-K
SIALW214000-10KG-KEA
514
EUR
SIALW214000-10KG-K
SIALW214000-1KG-K
SIALW214000-5KG-K
Benzylbutyrat, Sigma-Aldrich®
Benzylbutyrat
Formel:
C₁₁H₁₄O₂ Molecular Weight: 178.23 g/mol Siedepunkt: 240 Dichte: 1.01 g/cm³ (20 °C) |
MDL:
MFCD00027133 CAS-Nummer: 103-37-7 EINECS: 203-105-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...