Benzylbutyrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Butyric acid benzyl ester, Butanoic acid benzyl ester

M-Clarity™ Qualitätsstufe = MQ300

W214019-100G-K W214019-1KG-K
SIALW214019-100G-KEA 131 EUR
SIALW214019-100G-K SIALW214019-1KG-K
Benzylbutyrat, Sigma-Aldrich®
Benzylbutyrat
Formel: C₁₁H₁₄O₂
Molecular Weight: 178,23 g/mol
Siedepunkt: 240 °C (1013 hPa)
Dichte: 1,01 g/cm³ (20 °C)
MDL: MFCD00027133
CAS-Nummer: 103-37-7
EINECS: 203-105-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR