2-Phenylethylacetat ≥98%
Lieferant: Thermo Scientific
Synonyme:
Essigsäure(2-phenylethyl)ester, Phenethylacetat, (2-Phenylethyl)acetat
B21238.0B
B21238.30
B21238.0BEA
91
EUR
B21238.0B
B21238.30
2-Phenylethylacetat ≥98%
2-Phenylethylacetat
Purity: 98%
CAS No.: 103-45-7
Molecular Formula: CH3CO2(CH2)2C6H5
Formula Weight: 164.21
Flash Point: 107°(225°F)
Refractive Index: 1.4990
Odour: Fruit-like
Melting Point: -31°
Boiling Point: 231-233°
Beilstein Reference: 638179
MDL No.: MFCD00008720
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,2 g/mol Siedepunkt: 231…233 °C Schmelzpunkt: –31 °C Dichte: 1,029 Flammpunkt: 107 °C (225 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008720 CAS-Nummer: 103-45-7 EINECS: 203-113-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...