Dibenzylmalonat 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Malonic acid dibenzyl ester, 1,3-Propanedioic acid dibenzyl ester
A10844.09
A10844.18
A10844.30
A10844.09EA
52.9
EUR
A10844.09
A10844.18
A10844.30
Dibenzylmalonat 95%
Dibenzylmalonat
Purity: 95%
CAS No.: 15014-25-2
Molecular Formula: C17H16O4
Formula Weight: 284.31
Flash Point:
Formel:
C₁₇H₁₆O₄ Molecular Weight: 284,31 g/mol Siedepunkt: 188 °C (0,2 mmHg) Dichte: 1,137 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004779 CAS-Nummer: 15014-25-2 EINECS: 239-099-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...