Dibenzylmalonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Malonic acid dibenzyl ester, 1,3-Propanedioic acid dibenzyl ester
M-Clarity™ Qualitätsstufe = MQ100
160407-10G
160407-50G
SIAL160407-10GEA
67.5
EUR
SIAL160407-10G
SIAL160407-50G
Dibenzylmalonat, Sigma-Aldrich®
Dibenzylmalonat
Formel:
C₁₇H₁₆O₄ Molecular Weight: 284,31 g/mol Siedepunkt: 188 °C (1013 hPa) Dichte: 1,137 g/cm³ (20 °C) |
MDL:
MFCD00004779 CAS-Nummer: 15014-25-2 EINECS: 239-099-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...