8-Oxo-8-hydroadenosin ≥98,0% (durch HPLC)
Lieferant: TCI
Synonyme:
7,8-Dihydro-8-oxoadenosine, 8-Hydroxyadenosine
O0401-200MG
O0401-1G
TCIAO0401-200MGEA
175
EUR
TCIAO0401-200MG
TCIAO0401-1G
8-Oxo-8-hydroadenosin ≥98,0% (durch HPLC)
8-Oxo-8-hydroadenosin
Formel:
C₁₀H₁₃N₅O₅ Molecular Weight: 283.24 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD15145291 CAS-Nummer: 29851-57-8 |
|
Spezifikation Testergebnisse
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 23.50 to 25.30 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...