Ethylcinnamat 98% (trans-Isomere)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
150531000
150535000
150530025
ACRO150531000EA
24.8
EUR
ACRO150531000
ACRO150535000
ACRO150530025
Ethylcinnamat 98% (trans-Isomere)
Ethylcinnamat
Formel:
C₁₁H₁₂O₂ Siedepunkt: 271 °C (760 mmHg) Schmelzpunkt: 6,5…7.5 °C Dichte: 1,046 g/cm³ Flammpunkt: 135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009189 CAS-Nummer: 103-36-6 EINECS: 203-104-6 |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5580 to 1.5610 (20°C, 589 nm) |
GC | ≥97.5 % |
Specific gravity | (25/25°C) 1.045 to 1.048 |
Acid value | ≤1 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...