Ölrot O 0,5% in Propylenglykol, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
CI 26125
M-Clarity™ Qualitätsstufe = MQ200
O1516-250ML
O1516-500ML
O1516-250MLEA
367
EUR
O1516-250ML
O1516-500ML
Ölrot O 0,5% in Propylenglykol, Sigma-Aldrich®
Ölrot O
Oil red O is a lysochrome (fat-soluble dye) diazo dye used for staining of neutral triglycerides and lipids on frozen sections and some lipoproteins on paraffin sections. In histology, a supersaturated solution of oil red O in isopropanol may be used to stain fat in tissue.
Formel:
C₂₆H₂₄N₄O Molecular Weight: 408,51 g/mol Lagertemperatur: Room Temperature |
MDL:
MFCD00003898 CAS-Nummer: 1320-06-5 |
Spezifikation Testergebnisse
Appearance Dark Red, Sediment Free |
Conforms |
UV/VIS Absorbance Lambda Max 506 to 520 nm |
0.40 - 1.00 |
Lambda Max | 506 - 520 nm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...