4-(Tetrahydropyran-2-yloxy)phenylboronsäure

Lieferant: Thermo Scientific

Synonyme: 4-(2-Tetrahydropyranyloxy)benzeneboronic acid, 4-(Tetrahydro-2H-pyran-2-yloxy)phenylboronic acid, 4-(Tetrahydro-2H-pyran-2-yloxy)benzeneboronic acid, (4-((Tetrahydro-2H-pyran-2-yl)oxy)phenyl)boronic acid

AC35945.250 AC35945.1
MAYBAC35945.250EA 0 EUR
MAYBAC35945.250 MAYBAC35945.1
4-(Tetrahydropyran-2-yloxy)phenylboronsäure
4-(Tetrahydropyran-2-yloxy)phenylboronsäure
Formel: C₁₁H₁₅BO₄
MDL: MFCD02183525
CAS-Nummer: 182281-01-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR