2-Brom-4-chlor-6-fluoranilin
Lieferant: CHEMPUR
FL13189.POR
013189-25G
013189-5G
CHMPFL13189.POREA
0
EUR
CHMPFL13189.POR
FLUO013189-25G
FLUO013189-5G
2-Brom-4-chlor-6-fluoranilin
2-Brom-4-chlor-6-fluoranilin
Formel:
C₆H₄BrClFN Molecular Weight: 224.46 g/mol |
MDL:
MFCD00070746 CAS-Nummer: 195191-47-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...