2-Brom-4-(trifluormethoxy)anilin

Lieferant: Thermo Scientific

Synonyme: 2-Bromo-4-trifluoromethoxyaniline

TL00668.1 TL00668.10 TL00668.50
MAYBTL00668.1EA 0 EUR
MAYBTL00668.1 MAYBTL00668.10 MAYBTL00668.50
2-Brom-4-(trifluormethoxy)anilin
2-Brom-4-(trifluormethoxy)anilin
Formel: C₇H₅BrF₃NO
Molecular Weight: 256,02 g/mol
MDL: MFCD00203478
CAS-Nummer: 175278-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR