2-Brom-4-(trifluormethoxy)anilin
Lieferant: FLUOROCHEM
004298-25G
004298-100G
FLUO004298-25GEA
42.9
EUR
FLUO004298-25G
FLUO004298-100G
2-Brom-4-(trifluormethoxy)anilin
2-Brom-4-(trifluormethoxy)anilin
Formel:
C₇H₅BrF₃NO |
MDL:
MFCD00203478 CAS-Nummer: 175278-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...