4-Brom-2-(trifluormethoxy)anilin

Lieferant: FLUOROCHEM

004299-5G 004299-25G
FLUO004299-5GEA 0 EUR
FLUO004299-5G FLUO004299-25G
4-Brom-2-(trifluormethoxy)anilin
4-Brom-2-(trifluormethoxy)anilin
Formel: C₇H₅BrF₃NO
MDL: MFCD00179338
CAS-Nummer: 175278-09-8
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR