4-Brom-2-nitroanilin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
B25468.22
B25468.14
B25468.22EA
202
EUR
B25468.22
B25468.14
4-Brom-2-nitroanilin ≥98%
4-Brom-2-nitroanilin
Purity: 98%
CAS No.: 875-51-4
Molecular Formula: C6H5BrN2O2
Formula Weight: 217.02
Melting Point: 111-114°
MDL No.: MFCD00041312
Formel:
C₆H₅BrN₂O₂ Molecular Weight: 217,02 g/mol Siedepunkt: 309 °C (1013 hPa) Schmelzpunkt: 111…114 °C Dichte: 1,81 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041312 CAS-Nummer: 875-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...