2-Brom-6-nitrophenol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

643378-25G 643378-5G
SIAL643378-25GEA 441 EUR
SIAL643378-25G SIAL643378-5G
2-Brom-6-nitrophenol, Sigma-Aldrich®
2-Brom-6-nitrophenol
Formel: C₆H₄BrNO₃
Molecular Weight: 218,01 g/mol
MDL: MFCD02683216
CAS-Nummer: 13073-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR