4-Brom-2-methoxyphenol

Lieferant: CHEMPUR

Synonyme: 4-Bromguajacol, 1-Brom-4-hydroxy-3-methoxybenzol, 5-Brom-2-hydroxyanisol

BTB13905.250 BTB13905.1
CHMPBTB13905.250EA 0 EUR
CHMPBTB13905.250 MAYBBTB13905.1
4-Brom-2-methoxyphenol
4-Brom-2-methoxyphenol
Formel: C₇H₇BrO₂
Molecular Weight: 203.04 g/mol
MDL: MFCD00051937
CAS-Nummer: 7368-78-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR