2,6-Dichlor-4-(trifluormethoxy)anilin

Lieferant: FLUOROCHEM

007843-10G 007843-25G
FLUO007843-10GEA 140 EUR
FLUO007843-10G FLUO007843-25G
2,6-Dichlor-4-(trifluormethoxy)anilin
2,6-Dichlor-4-(trifluormethoxy)anilin
Formel: C₇H₄Cl₂F₃NO
Molecular Weight: 246.02 g/mol
MDL: MFCD00190128
CAS-Nummer: 99479-66-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR