2-Chlor-4-(trifluormethoxy)anilin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
655929-1G
655929-5G
SIAL655929-1GEA
26
EUR
SIAL655929-1G
SIAL655929-5G
2-Chlor-4-(trifluormethoxy)anilin, Sigma-Aldrich®
2-Chlor-4-(trifluormethoxy)anilin
Formel:
C₇H₅ClF₃NO Molecular Weight: 211,57 g/mol |
MDL:
MFCD01631704 CAS-Nummer: 69695-61-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...