4-Chlor-2-fluorphenol

Lieferant: CHEMPUR

FL13284.POR 013284-25G 013284-100G
CHMPFL13284.POREA 0 EUR
CHMPFL13284.POR FLUO013284-25G FLUO013284-100G
4-Chlor-2-fluorphenol
4-Chlor-2-fluorphenol
Formel: C₆H₄ClFO
Molecular Weight: 146.55 g/mol
MDL: MFCD00075030
CAS-Nummer: 348-62-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR