3-Chlor-5-fluorphenol ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

H26195.03 H26195.06
H26195.03EA 75.9 EUR
H26195.03 H26195.06
3-Chlor-5-fluorphenol ≥98%
3-Chlor-5-fluorphenol

Purity: 98% CAS No.: 202982-70-5 Molecular Formula: C6H4ClFO Formula Weight: 146.55 Refractive Index: 1.537

Formel: C₆H₄ClFO
Lagertemperatur: Raumtemperatur
MDL: MFCD00042212
CAS-Nummer: 202982-70-5
EINECS: None
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR