3-Chlor-5-fluorphenol

Lieferant: CHEMPUR

FL13283.POR 013283-1G 013283-5G
CHMPFL13283.POREA 0 EUR
CHMPFL13283.POR FLUO013283-1G FLUO013283-5G
3-Chlor-5-fluorphenol
3-Chlor-5-fluorphenol
Formel: C₆H₄ClFO
MDL: MFCD00042212
CAS-Nummer: 202982-70-5
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR