2-Chlor-4-iodanilin

Lieferant: Thermo Scientific

S01378.1 S01378.10 S01378.50
MAYBS01378.1EA 0 EUR
MAYBS01378.1 MAYBS01378.10 MAYBS01378.50
2-Chlor-4-iodanilin
2-Chlor-4-iodanilin
Formel: C₆H₅ClIN
Molecular Weight: 253,47 g/mol
Siedepunkt: 289 °C (1013 hPa)
Schmelzpunkt: 73 °C
MDL: MFCD00025298
CAS-Nummer: 42016-93-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR