4-Chlor-3-nitrophenylisothiocyanat

Lieferant: FLUOROCHEM

Synonyme: 1-Chloro-4-isothiocyanato-3-nitrobenzene

008772-5G 008772-10G
FLUO008772-5GEA 101 EUR
FLUO008772-5G FLUO008772-10G
4-Chlor-3-nitrophenylisothiocyanat
4-Chlor-3-nitrophenylisothiocyanat
Formel: C₇H₃ClN₂O₂S
Schmelzpunkt: 53…54 °C
MDL: MFCD00041236
CAS-Nummer: 127142-66-9
UN: 2923
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR