Methyl-2-isothiocyanatobenzoat

Lieferant: Thermo Scientific

Synonyme: o-Methoxycarbonylphenylisothiocyanat

SB01695.1 SB01695EA SB01695.50
MAYBSB01695.1EA 0 EUR
MAYBSB01695.1 MAYBSB01695EA MAYBSB01695.50
Methyl-2-isothiocyanatobenzoat
Methyl-2-isothiocyanatobenzoat
Formel: C₉H₇NO₂S
Molecular Weight: 193,23 g/mol
Siedepunkt: 104 °C (1013 hPa)
Dichte: 1,25 g/cm³ (20 °C)
MDL: MFCD00041062
CAS-Nummer: 16024-82-1
EINECS: 240-164-2
UN: 2923
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR