Dimethylisophthalat ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Isophthalsäuredimethylester
B23779.0E
B23779.22
B23779.36
B23779.0EEA
148
EUR
B23779.0E
B23779.22
B23779.36
Dimethylisophthalat ≥98%
Dimethylisophthalat
Purity: 98%
CAS No.: 1459-93-4
Molecular Formula: C10H10O4
Formula Weight: 194.19
Flash Point: 138°(280°F)
Odour: Mild
Boiling Point: 282°
Beilstein Reference: 1912251
MDL No.: MFCD00008433
Formel:
C₁₀H₁₀O₄ Molecular Weight: 194.19 g/mol Siedepunkt: 282 °C Schmelzpunkt: 67…70 °C Dichte: 1.194 Flammpunkt: 138 °C (280 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008433 CAS-Nummer: 1459-93-4 EINECS: 215-951-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...