5-Aminoisophthalsäure
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
5-Amino-1,3-benzoldicarbonsäure
RJC02328.250
RJC02328.1
CHMPRJC02328.250EA
0
EUR
CHMPRJC02328.250
MAYBRJC02328.1
5-Aminoisophthalsäure
5-Aminoisophthalsäure
Formel:
C₈H₇NO₄ Molecular Weight: 181,15 g/mol Siedepunkt: 515 °C (1013 hPa) Schmelzpunkt: 150…156 °C |
MDL:
MFCD00002522 CAS-Nummer: 99-31-0 EINECS: 202-748-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...