5-Nitroisophthalsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Nitro-1,3-benzoldicarbonsäure
A11696.22
A11696.36
A11696.22EA
29
EUR
A11696.22
A11696.36
5-Nitroisophthalsäure ≥98%
5-Nitroisophthalsäure
Purity: 98%
CAS No.: 618-88-2
Molecular Formula: C8H5NO6
Formula Weight: 211.13
Odour: Odourless
Melting Point: 260-264°
Beilstein Reference: 1976587
MDL No.: MFCD00007254
Formel:
C₈H₅NO₆ Schmelzpunkt: 260…264 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007254 CAS-Nummer: 618-88-2 EINECS: 210-568-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...