5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)
Lieferant: Uptima
|
Achtung
|
Synonyme:
Bis(4-nitrophenyl)disulfid-3,3'-dicarbonsäure, DTNB, 2,2'-Dinitro-5,5'-dithiodibenzoesäure, Ellman's Reagenz, 3,3'-Dithiobis(6-nitrobenzoesäure)
UP01566I
UP01566H
UPTIUP01566IEA
0
EUR
UPTIUP01566I
UPTIUP01566H
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)
Formel:
C₁₄H₈N₂O₈S₂ Molecular Weight: 396.36 g/mol Schmelzpunkt: 240…244 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007140 CAS-Nummer: 69-78-3 EINECS: 200-714-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...