5-Fluor-2-iodanilin ≥97%

Lieferant: Thermo Scientific
Gefahr

L17723.03 L17723.06
L17723.03EA 44.8 EUR
L17723.03 L17723.06
5-Fluor-2-iodanilin ≥97%
5-Fluor-2-iodanilin

Purity: 97% CAS No.: 255724-71-1 Molecular Formula: C6H5FIN Formula Weight: 237.02 Melting Point: 40-42° Beilstein Reference: 8478850 UN No.: 2811 MDL No.: MFCD02093953

Formel: C₆H₅FIN
Molecular Weight: 237,02 g/mol
Siedepunkt: 259 °C (1013 hPa)
Schmelzpunkt: 40…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093953
CAS-Nummer: 255724-71-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR