4-Fluor-2-nitrophenol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-Fluoro-4-hydroxy-3-nitrobenzene
A15847.06
A15847.14
A15847.06EA
26.8
EUR
A15847.06
A15847.14
4-Fluor-2-nitrophenol ≥98%
4-Fluor-2-nitrophenol
Purity: 98+%
CAS No.: 394-33-2
Molecular Formula: C6H4FNO3
Formula Weight: 157.10
Melting Point: 73-76°
Beilstein Reference: 1950412
Formel:
C₆H₄FNO₃ Molecular Weight: 157,1 g/mol Schmelzpunkt: 73…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042528 CAS-Nummer: 394-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...