4-Fluor-2-nitrophenol

Lieferant: Thermo Scientific

XAX00012.1 XAX00012.10 XAX00012.50
MAYBXAX00012.1EA 0 EUR
MAYBXAX00012.1 MAYBXAX00012.10 MAYBXAX00012.50
4-Fluor-2-nitrophenol
4-Fluor-2-nitrophenol
Formel: C₆H₄FNO₃
Schmelzpunkt: 74 °C
MDL: MFCD00042528
CAS-Nummer: 394-33-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR