4-Fluor-2,6-dinitrophenol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
A17309.03
A17309.06
A17309.03EA
31.2
EUR
A17309.03
A17309.06
4-Fluor-2,6-dinitrophenol ≥98%
4-Fluor-2,6-dinitrophenol
Purity: 98%
CAS No.: 364-32-9
Molecular Formula: C6H3FN2O5
Formula Weight: 202.10
Melting Point: 50-56°
MDL No.: MFCD00179333
Formel:
C₆H₃FN₂O₅ Molecular Weight: 202,1 g/mol Schmelzpunkt: 50…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00179333 CAS-Nummer: 364-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...