4-Nitro-o-anisidin ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Methoxy-4-nitroanilin, 2-Amino-5-nitroanisol
B24220.14
B24220.22
B24220.36
B24220.14EA
23.9
EUR
B24220.14
B24220.22
B24220.36
4-Nitro-o-anisidin ≥99%
4-Nitro-o-anisidin
Purity: 99%
CAS No.: 97-52-9
Molecular Formula: C7H8N2O3
Formula Weight: 168.15
Melting Point: 138-142°
Beilstein Reference: 879619
Formel:
C₇H₈N₂O₃ Molecular Weight: 168,15 g/mol Siedepunkt: 364 °C (1013 hPa) Schmelzpunkt: 138…142 °C Dichte: 1,211 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007363 CAS-Nummer: 97-52-9 EINECS: 202-588-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...