5-Methoxy-2-nitrophenol 98%
Lieferant: Thermo Scientific
|
Gefahr
|
335440010
335440050
ACRO335440010EA
104
EUR
ACRO335440010
ACRO335440050
5-Methoxy-2-nitrophenol 98%
5-Methoxy-2-nitrophenol
Formel:
C₇H₇NO₄ Molecular Weight: 169,14 g/mol Schmelzpunkt: 90…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00100932 CAS-Nummer: 704-14-3 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Yellow Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 90 to 94 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...