5-Methoxy-2-nitrophenol

Lieferant: CHEMPUR

NRB01450.250 NRB01450.1
CHMPNRB01450.250EA 0 EUR
CHMPNRB01450.250 MAYBNRB01450.1
5-Methoxy-2-nitrophenol
5-Methoxy-2-nitrophenol
Formel: C₇H₇NO₄
Molecular Weight: 169.14 g/mol
Schmelzpunkt: 95
MDL: MFCD00100932
CAS-Nummer: 704-14-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR