(Methylsulfonyl)benzol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methylphenylsulfon
A11727.09
A11727.18
A11727.09EA
48.8
EUR
A11727.09
A11727.18
(Methylsulfonyl)benzol ≥98%
(Methylsulphonyl)benzene
Purity: 98+%
CAS No.: 3112-85-4
Molecular Formula: C7H8O2S
Formula Weight: 156.20
Melting Point: 85-88°
Beilstein Reference: 1906914
Formel:
C₇H₈O₂S Molecular Weight: 156,21 g/mol Schmelzpunkt: 85…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014741 CAS-Nummer: 3112-85-4 EINECS: 221-476-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...