(Methylsulfonyl)benzol

Lieferant: FLUOROCHEM

001313-10G 001313-25G
FLUO001313-10GEA 31.1 EUR
FLUO001313-10G FLUO001313-25G
(Methylsulfonyl)benzol
(Methylsulphonyl)benzene
Formel: C₇H₈O₂S
MDL: MFCD00014741
CAS-Nummer: 3112-85-4
EINECS: 221-476-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR